Chlorobis(4-fluorophenyl)phosphine ≥98%

Leverandør: Thermo Scientific
Danger

H26953.03
H26953.03EA 4475 NOK
H26953.03
Chlorobis(4-fluorophenyl)phosphine ≥98%
Chlorobis(4-fluorophenyl)phosphine

Purity: 98% CAS No.: 23039-97-6 Molecular Formula: C12H8ClF2P Formula Weight: 256.62 UN No.: 3265 MDL No.: MFCD04972302

Formel: C₁₂H₈ClF₂P
MW: 256,62 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD04972302
CAS-nummer: 23039-97-6
FN: 3265
ADR: 8,II

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR