cis-4-Hydroxy-D(+)-proline ≥98%

Leverandør: Thermo Fisher Scientific

Synonymer: (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline

H27633.03
H27633.03EA 1475 NOK
H27633.03
cis-4-Hydroxy-D(+)-proline ≥98%
cis-4-Hydroxy-D(+)-proline

Purity: 98+% CAS No.: 2584-71-6 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: 243° dec. MDL No.: MFCD00005252

Formel: C₅H₉NO₃
MW: 131.13 g/mol
Smeltepunkt: 243 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005252
CAS-nummer: 2584-71-6
EINECS: 219-963-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR