6-Cyanobenzothiazole 98%

Leverandør: Thermo Fisher Scientific
Danger

H32454.06
H32454.06EA 5400 NOK
H32454.06
6-Cyanobenzothiazole 98%
6-Cyanobenzothiazole

Purity: 98% CAS No.: 58249-61-9 Molecular Formula: C8H4N2S Formula Weight: 160.20 Melting Point: 137-138° UN No.: UN3439 MDL No.: MFCD06659656

Formel: C₈H₄N₂S
Smeltepunkt: 137…138 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06659656
CAS-nummer: 58249-61-9
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR