Palladium 5% on carbon, pulver Type 490

Leverandør: Thermo Fisher Scientific
Warning

H33217.09 H33217.18
H33217.09EA 13075 NOK
H33217.09 H33217.18
Palladium 5% on carbon, pulver Type 490
Palladium
Formel: Pd
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011167
CAS-nummer: 7440-05-3
EINECS: 231-115-6
FN: 1325
ADR: 4.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR