6-Chloroimidazo[1,2-a]pyridine ≥97%

Leverandør: Thermo Fisher Scientific
Warning

H33234.03 H33234.06
H33234.03EA 390 NOK
H33234.03 H33234.06
6-Chloroimidazo[1,2-a]pyridine ≥97%
6-Chloroimidazo[1,2-a]pyridine
Formel: C₇H₅ClN₂
Smeltepunkt: 80…82 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05663807
CAS-nummer: 6188-25-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR