Rhodamine 6G perchlorate 96%

Leverandør: Thermo Fisher Scientific
Warning

H33626.MD H33626.03
H33626.MDEA 540 NOK
H33626.MD H33626.03
Rhodamine 6G perchlorate 96%
Rhodamine 6G perchlorate

Purity: 96% CAS No.: 13161-28-9 Molecular Formula: C28H31ClN2O7 Formula Weight: 543.02 Melting Point: 240° MDL No.: MFCD00013117

Formel: C₂₈H₃₁ClN₂O₇
Smeltepunkt: >240 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013117
CAS-nummer: 13161-28-9
EINECS: 236-099-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR