3-(4-n-Propoxyphenyl)propionic acid 96%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 3-(4-Propoxyphenyl)propanoic acid

H33646.03 H33646.06
H33646.03EA 600 NOK
H33646.03 H33646.06
3-(4-n-Propoxyphenyl)propionic acid 96%
3-(4-n-Propoxyphenyl)propionic acid

Purity: 96% CAS No.: 3243-40-1 Molecular Formula: C12H16O3 Formula Weight: 208.26 Melting Point: 94-99° MDL No.: MFCD00484121

Formel: C₁₂H₁₆O₃
MW: 208.26 g/mol
Smeltepunkt: 94…99 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00484121
CAS-nummer: 3243-40-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR