Diethyl chlorophosphate ≥97%

Leverandør: Thermo Fisher Scientific
Danger

H33820.14 H33820.22
H33820.14EA 610 NOK
H33820.14 H33820.22
Diethyl chlorophosphate ≥97%
Diethyl chlorophosphate
Formel: C₄H₁₀ClO₃P
Kokepunkt: 60 °C (2 mmHg)
Tetthet: 1,194
Flash punkt: 61 °C (142 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009075
CAS-nummer: 814-49-3
EINECS: 212-369-4
FN: 3278
ADR: 6.1,I

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR