3-Iodochromone ≥97%

Leverandør: Thermo Fisher Scientific
Danger

H34295.MD H34295.03
H34295.MDEA 1625 NOK
H34295.MD H34295.03
3-Iodochromone ≥97%
3-Iodochromone
Formel: C₉H₅IO₂
Smeltepunkt: 95…97 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09475514
CAS-nummer: 122775-34-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR