7-Bromoimidazo[1,2-a]pyridine 95%

Leverandør: Thermo Fisher Scientific
Warning

H35721.03
H35721.03EA 2275 NOK
H35721.03
7-Bromoimidazo[1,2-a]pyridine 95%
7-Bromoimidazo[1,2-a]pyridine
Formel: C₇H₅BrN₂
Smeltepunkt: 118…120 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD07778353
CAS-nummer: 808744-34-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR