6-Fluoroquinoline ≥97%

Leverandør: Thermo Fisher Scientific
Warning

H35900.MD H35900.03
H35900.MDEA 730 NOK
H35900.MD H35900.03
6-Fluoroquinoline ≥97%
6-Fluoroquinoline
Formel: C₉H₆FN
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01685512
CAS-nummer: 396-30-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR