Palladium 5% on calcium carbonate lead poisoned A305060-5

Leverandør: Thermo Fisher Scientific
Danger

H36186.06
H36186.06EA 2325 NOK
H36186.06
Palladium 5% on calcium carbonate lead poisoned A305060-5
Palladium

Lindlar hydrogenation catalyst


CAS No.: 7440-05-3 Physical Form: Powder Appearance: Dark grey Odour: Odourless MDL No.: MFCD03427452

Formel: Pd
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03427452
CAS-nummer: 7440-05-3
EINECS: 231-115-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR