2-Fluoro-3-iodopyridine ≥97%

Leverandør: Thermo Fisher Scientific
Warning

H50146.03
H50146.03EA 3400 NOK
H50146.03
2-Fluoro-3-iodopyridine ≥97%
2-Fluoro-3-iodopyridine

Purity: 97% CAS No.: 113975-22-7 Molecular Formula: C5H3FIN Formula Weight: 222.99 MDL No.: MFCD03095287

Formel: C₅H₃FIN
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03095287
CAS-nummer: 113975-22-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR