N-Benzhydryl-N-methylamine 98%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: N-Methylbenzhydrylamine

H50327.03 H50327.06
H50327.03EA 645 NOK
H50327.03 H50327.06
N-Benzhydryl-N-methylamine 98%
N-Benzhydryl-N-methylamine

Purity: 98% CAS No.: 14683-47-7 Molecular Formula: C14H15N Formula Weight: 197.28 Melting Point: 38-42° Beilstein Reference: 2096409 MDL No.: MFCD00467853

Formel: C₁₄H₁₅N
MW: 197.28 g/mol
Smeltepunkt: 38…42 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00467853
CAS-nummer: 14683-47-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR