anti-(1R)-(+)-Camphorquinone 3-oxime 99%

Leverandør: Thermo Fisher Scientific

Synonymer: anti-(1R)-(+)-Camphorquinone 3-Oxime, (1R,E)-(+)-2,3-Bornanedione 3-oxime,anti-(1R)-(+)-Camphorquinone 3-oxime, anti-(1R)-(+)-2,3-Bornanedione 3-Oxime

H51034.06
H51034.06EA 3750 NOK
H51034.06
anti-(1R)-(+)-Camphorquinone 3-oxime 99%
anti-(1R)-(+)-Camphorquinone 3-oxime

Purity: 99% CAS No.: 31571-14-9 Molecular Formula: C10H15NO2 Formula Weight: 181.23 Beilstein Reference: 3200377 MDL No.: MFCD00074848

Formel: C₁₀H₁₅NO₂
MW: 181.24 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00074848
CAS-nummer: 31571-14-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR