(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid ≥98%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: (1R,2R)-Cyclohexane-1,2-dicarboxylic acid, (1R,2R)-(-)-trans-Cyclohexane-1,2-dicarboxylic acid

H52293.MD H52293.03 H52293.06
H52293.MDEA 555 NOK
H52293.MD H52293.03 H52293.06
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid ≥98%
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid

Purity: 98+% CAS No.: 46022-05-3 Molecular Formula: C8H12O4 Formula Weight: 172.18 Physical Form: Solid Appearance: White MDL No.: MFCD00062973

Formel: C₈H₁₂O₄
MW: 172.18 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00062973
CAS-nummer: 46022-05-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR