N4-Benzoyl-2'-deoxycytidine ≥98%

Leverandør: Thermo Fisher Scientific

H52298.03 H52298.06
H52298.03EA 1575 NOK
H52298.03 H52298.06
N4-Benzoyl-2'-deoxycytidine ≥98%
N4-Benzoyl-2'-deoxycytidine

Purity: 98+% CAS No.: 4836-13-9 Molecular Formula: C16H17N3O5 Formula Weight: 331.33 MDL No.: MFCD00010115

Formel: C₁₆H₁₇N₃O₅
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010115
CAS-nummer: 4836-13-9
EINECS: 225-421-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR