Boc-aza-DL-leucine 97%

Leverandør: Thermo Fisher Scientific

H52412.03 H52412.06
H52412.03EA 1575 NOK
H52412.03 H52412.06
Boc-aza-DL-leucine 97%
Boc-aza-DL-leucine

Purity: 97% CAS No.: 851653-36-6 Molecular Formula: C10H20N2O4 Formula Weight: 232.28 MDL No.: MFCD04972270

Formel: C₁₀H₂₀N₂O₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD04972270
CAS-nummer: 851653-36-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR