Boc-D-α-methylserine 97%

Leverandør: Thermo Fisher Scientific

H52787.03
H52787.03EA 10650 NOK
H52787.03
Boc-D-α-methylserine 97%
Boc-D-alpha-methylserine

Purity: 97% CAS No.: 84311-18-2 Molecular Formula: C9H17NO5 Formula Weight: 219.24 MDL No.: MFCD02682596

Formel: C₉H₁₇NO₅
MW: 219.24 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02682596
CAS-nummer: 84311-18-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR