6-Bromooxindole ≥97%

Leverandør: Thermo Fisher Scientific

H56012.03 H56012.06
H56012.03EA 970 NOK
H56012.03 H56012.06
6-Bromooxindole ≥97%
6-Bromooxindole
Formel: C₈H₆BrNO
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02179605
CAS-nummer: 99365-40-9

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR