Azetidine ≥98%

Leverandør: Thermo Fisher Scientific
Danger

H56351.06
H56351.06EA 8450 NOK
H56351.06
Azetidine ≥98%
Azetidine
Formel: C₃H₇N
Kokepunkt: 61…62 °C
Tetthet: 0,847
Flash punkt: –21 °C (–6 °F)
MDL nummer: MFCD00005165
CAS-nummer: 503-29-7
EINECS: 207-963-8
ADR: 3,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR