3-Iodooxetane 95%

Leverandør: Thermo Fisher Scientific
Warning

H57302.MDEA 2975 NOK
H57302.MD H57302.ME
3-Iodooxetane 95%
3-Iodooxetane
Formel: C₃H₅IO
Kokepunkt: 166 °C
Tetthet: 2,14
Flash punkt: 54 °C (129 °F)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD08544403
CAS-nummer: 26272-85-5
FN: 1993
ADR: 3,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR