D-β-Proline ethyl ester hydrochloride 97%
Leverandør: Thermo Fisher Scientific
Warning
H57789.03
H57789.03EA 1975 NOK
H57789.03
D-β-Proline ethyl ester hydrochloride 97%
D-beta-Proline ethyl ester hydrochloride
Purity: 97% Molecular Formula: C7H13NO2.HCl Formula Weight: 179.65 Storage Temperature: 2°C to 8°C Optical Rotation: -14° (c=0.6 in water) MDL No.: MFCD18071037