β-Carotene 99%

Leverandør: Thermo Fisher Scientific

Synonymer: 2,2'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-Tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(1,3,3-trimethylcyclohex-1-ene), Provitamin A

H60106.03 H60106.06
H60106.03EA 665 NOK
H60106.03 H60106.06
β-Carotene 99%
beta-Carotene

Purity: 99% CAS No.: 7235-40-7 Molecular Formula: C40H56 Formula Weight: 536.87 Storage Temperature: -30°C to -10°C Flash Point: 103°(217°F) Physical Form: Solid Density: 1.000 Sensitivity: Air and Light Sensitive Melting Point: 178-179° Beilstein Reference: 1917416 MDL No.: MFCD00001556

Formel: C₄₀H₅₆
MW: 536.89 g/mol
Smeltepunkt: 178…179 °C
Tetthet: 1.000
Flash punkt: 103 °C (217 °F)
MDL nummer: MFCD00001556
CAS-nummer: 7235-40-7
EINECS: 230-636-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR