(S)-(-)-β-Citronellol ≥97%

Leverandør: Thermo Fisher Scientific
Warning

H60174.03 H60174.06
H60174.03EA 690 NOK
H60174.03 H60174.06
(S)-(-)-β-Citronellol ≥97%
(S)-(-)-β-Citronellol
Formel: C₁₀H₂₀O
Kokepunkt: 225…226 °C
Tetthet: 0,856
Flash punkt: 99 °C (210 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00063214
CAS-nummer: 7540-51-4
EINECS: 231-415-7

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR