2-Amino-6-chloropyrazine 95%

Leverandør: Thermo Fisher Scientific
Warning

H61934.22 H61934.14
H61934.22EA 4475 NOK
H61934.22 H61934.14
2-Amino-6-chloropyrazine 95%
2-Amino-6-chloropyrazine
Formel: C₄H₄ClN₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00055024
CAS-nummer: 33332-28-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR