5-Bromocytosine 98%

Leverandør: Thermo Fisher Scientific
Danger

H61952.03
H61952.03EA 845 NOK
H61952.03
5-Bromocytosine 98%
5-Bromocytosine
Formel: C₄H₄BrN₃O
Smeltepunkt: 240…243 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00056025
CAS-nummer: 2240-25-7
EINECS: 218-806-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR