2-Amino-4-fluorobenzeneboronic acid pinacol ester 96%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Benzenamine, 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 2-Amino-4-fluorophenylboronic acid pinacol ester, 5-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

H62511.MD H62511.03
H62511.MDEA 2825 NOK
H62511.MD H62511.03
2-Amino-4-fluorobenzeneboronic acid pinacol ester 96%
2-Amino-4-fluorobenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C12H17BFNO2 Formula Weight: 237.08 Physical Form: Solid

Formel: C₁₂H₁₇BFNO₂
MW: 237.08 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD12025504
CAS-nummer: 1309982-16-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR