2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester 96%
Leverandør: Thermo Fisher Scientific
Synonymer: 2-Carbamoyl-3-fluorophenylboronic acid pinacol ester
H62904.MD H62904.03
H62904.MDEA 2450 NOK
H62904.MD H62904.03
2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester 96%
2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester
Purity: 96% Molecular Formula: C13H17BFNO3 Formula Weight: 265.09 Physical Form: Solid