Supelco® Organic Reference Standard, Vitamin B₁₂ₐ hydrochloride

Leverandør: Merck

H7126-100MG H7126-1G
H7126-100MGEA 1050 NOK
H7126-100MG H7126-1G
Supelco® Organic Reference Standard, Vitamin B₁₂ₐ hydrochloride
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c03cd860607263bfbc4665a5c99adeeaaebbfa54&vid=c03cd860607263bfbc4665a5c99adeeaaebbfa54&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b23dd41ae7c90f10ce9d6fed48919fd4bd59637&vid=0b23dd41ae7c90f10ce9d6fed48919fd4bd59637&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) Hydroxocobalamin hydrochloride
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 59461-30-2
MDL No. MFCD00167463
Molekylvekt 1382,82
Fraktforhold STD
Lagringsforhold 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR