Octadecyl derivatized silica 15-40 μm mean particle size, average diameter 60 Å, Supelco®

Leverandør: Merck
Warning

Synonymer: Derivatized Silica gel

H8261-100G
H8261-100GEA 9500 NOK
H8261-100G
Octadecyl derivatized silica 15-40 μm mean particle size, average diameter 60 Å, Supelco®
Octadecyl derivatized silica
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00166485
CAS-nummer: 66369-14-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f33506ea0e9d547e079bf3d29585211d80e20a9&vid=c88f71efdada024ab62ddb4d3771c994f31f8aed&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR