Stand for Imhoff sedimentation cones
Leverandør: Hach
206801
HACH206801EA 1275 NOK
HACH206801
Stand for Imhoff sedimentation cones
Stativer
Heavy gauge steel plated.
This stand is constructed to provide minimum interference with scale reading.