Settlometer kit
Leverandør: Hach
2115700
HACH2115700EA 3646.2 NOK
HACH2115700
Settlometer kit
Trakter Immhoff-trakter
This kit is used for analyzing activated sludge process. Custom PP paddle allows agitation of sample without scratching the jar.
  • Easy-to-read white graduations
  • Impact-resistant
  • Chemically resistant at pH range 3 to 10
Leveringsinformasjon: Kit includes a transparent PC 2 liter jar with cover and 10 PC graduated centrifuge tubes (15 ml).