1-(2-Pyridylazo)-2-naphthol (PAN) løsning 0,3% indicator for nickel and cobalt

Leverandør: Hach
Danger

Synonymer: PAN, 1-(2-Pyridylazo)-2-naphthol

2150232
HACH2150232EA 1050 NOK
HACH2150232
1-(2-Pyridylazo)-2-naphthol (PAN) løsning 0,3% indicator for nickel and cobalt
1-(2-Pyridylazo)-2-naphthol

For determination of low level nickel and cobalt by the PAN method.

Formel: C₁₅H₁₁N₃O
MW: 249,27 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004071
CAS-nummer: 85-85-8
FN: 3082
ADR: 9,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR