Filling Solution, Reference, 1 M KNO₃
Leverandør: Hach
28564026
HACH28564026EA 345 NOK
HACH28564026
Filling Solution, Reference, 1 M KNO₃
Elektrolytter Løsninger, elektroder
Filling solution for electrode reference element, potassium nitrate (KNO₃), 1 M solution.