BOD seed inoculum, POLYSEED
Leverandør: Hach
2918700
HACH2918700EA 4575 NOK
HACH2918700
BOD seed inoculum, POLYSEED
Mikrobiologiske testsystemer Tester for mikroorganismer
BOD seed inoculum for use in biochemical oxygen demand (BOD5) test procedures. A blend of specialised microbial cultures in an easy-to-use capsule. Designed to provide a uniform standard for the degradation of both industrial and municipal waste in BOD5 analyses.
Bestillingsinformasjon: Package of 50 capsules. International Laboratory Supply (InterLab), LTD.