Electrode storage bottles
Leverandør: Hach
9260300
HACH9260300EA 73 NOK
HACH9260300
Electrode storage bottles
Elektrodestativ
Electrode storage bottle is used to store pH electrodes.
  • Rubber grommet in lid ensures tight seal with 12 mm electrodes