Electrode cleaning solution for proteins/organic samples
Leverandør: Hach
C20C370
HACHC20C370EA 720 NOK
HACHC20C370
Electrode cleaning solution for proteins/organic samples
Elektrolytter Løsninger, lagring av elektroder
Electrode cleaning solution to remove proteins residues organic contaminations/deposits such as food stuff from pH sensing element.
Formula: PEPSIN in HCl