pH electrode, Radiometer Analytical
Leverandør: Hach
E11M001
HACHE11M001EA 3075 NOK
HACHE11M001
pH electrode, Radiometer Analytical
Elektroder pH-elektroder
These glass electrodes are compatible with Hach sensION+ benchtop pH meters (or any meter with a BNC connection). The PHG301 pH electrode should be used with Ag/AgCl or calomel reference electrodes.
  • Made with highly shock-resistant glass, making them extremely tough
  • Average lifetime of a correctly maintained Radiometer glass pH electrode is approximately two years
The PHG301 is a screw cap electrode and requires the CL114 cable for connection to a meter.
Leveringsinformasjon: Supplied with certificate of conformity.