Single-Port Portable Parallel Analyser (PPA), SL250
Leverandør: Hach
LPV4439911002
HACHLPV4439911002EA 34175 NOK
HACHLPV4439911002
Single-Port Portable Parallel Analyser (PPA), SL250
Kolorimeter Multiparameter Kolorimeter
One portable instrument - all the key parameters.
  • Probe-based tests include pH, dissolved oxygen, conductivity, and more
  • 5-Pin input connector: Two M12 connectors for IntelliCal™ probes
  • 130,8×58,9×258,3 mm dimension
Hach's SL250 Portable parallel analyser (PPA) is an innovative multiparameter water quality meter that allows you to simultaneously perform colorimetric and probe-based measurements. The instrument and corresponding Chemkey reagents are easy to use regardless of technical expertise. Compared to traditional powder or liquid reagent methods,More Product Information