Handheld TSS analyser
Leverandør: Hach
LXV3229900001
HACHLXV3229900001EA 0 NOK
HACHLXV3229900001
Handheld TSS analyser
Turbiditetsmålere
This Turbidity Suspended Solids (TSS) and sludge blanket level system can be used as an optimisation tool for monitoring process analyses or as an easy way to calibrate or validate online sensors.
  • Air bubble compensation for accuracy
  • Easy sludge blanket levels
  • Single, interval or continuous mode of operation
Combined multi-beam alternating light method with infrared diode system. It stores up to four different calibration curves for suspended solids and one for turbidity.
The probe is made of polished stainless steel with a scratch resistantMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Display Alphanumerical display, 4 rows of 16 characters
Minne
Oppløsning TSS: 0,001 (0 - 0,999) ; 0,01 (1 - 9,999) ; 0,1 (10 - 99,9) ; 1 (over 100) g/l
Turbidity: 0,001 (0 - 0,999) ; 0,01 (1 - 9,99) ; 0,1 (10 - 99,9) ; 1 (over 100 NTU) NTU
B×D×H 230×40×110 mm