Rack for cuvettes
Leverandør: Hach
LYW915
HACHLYW915EA 435 NOK
HACHLYW915
Rack for cuvettes
Stativer for rør Kyvettestativer
Rack made of PP, with numbered positions for 2×8 cuvettes of 10 mm pathlength.
  • Numbered positions to aid sample identification
  • Autoclavable at 121 °C