Multi-parameter electrode (pH/ORP/temperature), sensION+ 5045
Leverandør: Hach
LZW5045970002
HACHLZW5045970002EA 5750 NOK
HACHLZW5045970002
Multi-parameter electrode (pH/ORP/temperature), sensION+ 5045
Elektroder Elektroder med flere parametre
sensION+ 5045 is a combination multi-parameter electrode for determination of pH, ORP and temperature. Dedicated for use with Hach sensION+ MM110 portable meters.
  • Ceramic pin reference junction and glass (pH); platinum wire (ORP); and Pt1000 (temperature) sensors
  • Plastic body, non refillable gel reference electrolyte and built-in temperature sensor
  • Low maintenance design for a variety of applications
  • Ultra portable but protected against harsh field conditions
  • Heavy-duty electrode handle design optimised for field calibration and storage
  • Innovative designs