Multi-parameter electrode (pH/ORP/conductivity), sensION+ 5048
Leverandør: Hach
LZW5048970002
HACHLZW5048970002EA 7450 NOK
HACHLZW5048970002
Multi-parameter electrode (pH/ORP/conductivity), sensION+ 5048
Elektroder Elektroder med flere parametre
Multisensor for pH, ORP, conductivity and temperature. For use with sensION+ MM150 multi-parameter meter.
  • Accurate
  • Easy to use
  • Rugged and durable
  • Automatic temperature compensation
  • Glass (pH); platinum (conductivity & ORP); and Pt1000 (temperature) sensors
Attachable calibration tubes can screw into handle of electrode (only 2 ml of buffer or standard). Storage protector prevents breakage and avoids over pressurisation of the diaphragm. Ideal for pH, conductivity, ORP and temperature measurements in general aqueous applications.