ORP electrode, sensION+ 5056
Leverandør: Hach
LZW5056970002
HACHLZW5056970002EA 4675 NOK
HACHLZW5056970002
ORP electrode, sensION+ 5056
Elektroder Redox Electrodes
sensION+ 5056 is a combination platinum ORP electrode with a glass body and non refillable gel-electrolyte reference. It is dedicated for use with Hach sensION+ labpratory pH (mV) meters.
  • Fixed 1 meter cable and BNC connector
  • Low-maintenance design for a variety of applications
  • Gel-electrolyte reference and platinum wire sensor provides accurate, stable results over a wide range of applications
  • Encapsulated cartridge reference system ensures premium stability and long lifespan
  • Innovative reference system utilises a “cartridge” of Ag/AgCl crystals