Conductivity cell, sensION™ + 5062
Leverandør: Hach
LZW5062970002
HACHLZW5062970002EA 5025 NOK
HACHLZW5062970002
Conductivity cell, sensION™ + 5062
Elektroder Konduktivitetselektrode
The 5062 is ideal for conductivity measurements in extreme conditions and for 'difficult' applications, including dirty and viscous samples. Dedicated for use with Hach sensION+ portable conductivity meters.
  • Robust titanium cell provides reliable performance in extreme conditions
  • Ultra portable but protected against harsh field conditions
  • Heavy-duty electrode handle design optimized for field calibration and storage