Conductivity cell, Platinum sensION+ 5070
Leverandør: Hach
LZW5070970002
HACHLZW5070970002EA 4495.8 NOK
HACHLZW5070970002
Conductivity cell, Platinum sensION+ 5070
Elektroder Konduktivitetselektrode
This three-pole platinum cell with an integrated Pt1000 temperature sensor provides accurate, stable results over a wide measurement range.
  • Low maintenance requirements
  • Fixed 1 meter cable and telephonic connector
  • Dedicated for use with Hach sensION+ laboratory conductivity meters
  • Ideal for conductivity measurements in general aqueous applications
Leveringsinformasjon: Supplied with basic user manual.