Ion selective electrode, sodium, sensION+ 9650
Leverandør: Hach
LZW9650970002
HACHLZW9650970002EA 13975 NOK
HACHLZW9650970002
Ion selective electrode, sodium, sensION+ 9650
Elektroder ISE elektroder
sensION+ 9650 is a half-cell sodium ion selective electrode (ISE) with glass body and glass membrane selective to sodium ions in solution. It is recommended for use with the sensION+ 5044 reference electrode.
  • With fixed 1 meter cable and BNC connector
  • For use with a reference electorode
  • Provides fast, stable, and accurate response in a variety of sample types with low maintenance
This electrode is ideal for measuring sodium concentrations in wastewater, drinking water and general water quality applications.
Leveringsinformasjon: Supplied with basic user manual.