Syringe rack
Leverandør: HAMILTON BONADUZ
204880
HAMI204880EA 4850 NOK
HAMI204880
Syringe rack
Stativer for rør Syringe Racks
This brushed stainless steel rack is used for clean and safe storage of syringes with plastic holder. A lip on the plastic holder protects the syringe against accidental touching and falling down. The distance of the plastic holder prevents the syringe from getting in contact with the metal plate.
  • A trough underneath prevents the soiling of worplace with liquid
  • Usable as free standing or wall model