Micro syringe pipette
Leverandør: HAMILTON BONADUZ
84250
HAMI84250EA 2400 NOK
HAMI84250
Micro syringe pipette
Sprøyter Sprøyter for kromatografi
Microsyringe pipettes with a special ³/₄" electrotapered needle for easy attachment of PTFE tips. The sample only wets the disposable PTFE tips, eliminating sample cross-contamination.
  • The spring-loaded plunger and adjustable stops ensure excellent reproducibility
  • An adjustable stop allows the transfer of samples from 0,2 to 3 µl